Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzaldehyde, 97%

$181.64 - $181.64

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzaldehyde
CAS 188815-30-7
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD00061312
Beilstein 9121754
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2038506

 
alfa aesar™
B2038506
5g Each for $181.64
Description & Specifications

Specifications

Formula Weight 192.11
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C8H4F4O
MDL Number MFCD00061312
CAS 188815-30-7
Beilstein 9121754
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-tolualdehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 70°C (158°F)
Boiling Point 175°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.