missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-nitrobenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H63295MD

 View more versions of this product

Catalog No. AAH63295MD



Specifications

Specifications

3-Fluoro-5-nitrobenzoic acid
14027-75-9
MFCD05864360
VIPUIECMSDQUIK-UHFFFAOYSA-N
185.11
185.11
97%
250mg
C7H4FNO4
3-fluoro-5-nitrobenzoic acid, benzoic acid, 3-fluoro-5-nitro, 5-fluoro-3-nitrobenzoic acid, 3-fluoro-5-nitrobenzoic acid, pubchem1400, intermediates-zcf02073, spec-inf-bb s2256
C1=C(C=C(C=C1[N+](=O)[O-])F)C(=O)O
2759047
Solid

Chemical Identifiers

14027-75-9
185.11
VIPUIECMSDQUIK-UHFFFAOYSA-N
2759047
C7H4FNO4
MFCD05864360
3-fluoro-5-nitrobenzoic acid, benzoic acid, 3-fluoro-5-nitro, 5-fluoro-3-nitrobenzoic acid, 3-fluoro-5-nitrobenzoic acid, pubchem1400, intermediates-zcf02073, spec-inf-bb s2256
C1=C(C=C(C=C1[N+](=O)[O-])F)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents