missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53149MD

 View more versions of this product

Catalog No. AAH53149MD



Description

Description

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid
871332-63-7
MFCD07363793
CDJLVNMKXZMCHU-UHFFFAOYSA-N
[3-fluoro-5-(methylcarbamoyl)phenyl]boronic acid
44886927
98%
250mg
C8H9BFNO3
3-fluoro-5-methylcarbamoyl phenylboronic acid, 3-fluoro-5-methylcarbamoyl phenyl boronic acid, 3-fluoro-5-methylcarbamoyl benzeneboronic acid, n-methyl 3-borono-5-fluorobenzamide, 3-fluoro-5-methylaminocarbonyl phenylboronic acid, 3-fluoro-5-methylcarbamoyl phenyl boronic acid, acmc-209qgt, 3-fluoro-5-methylcarbamoyl benzene boronic acid, 3-fluoro-5-methylcarbamoyl-phenyl boronic acid, 3-fluoro-5-methyl carbamoyl phenylboronic acid
B(C1=CC(=CC(=C1)F)C(=O)NC)(O)O
196.972
196.97

Chemical Identifiers

871332-63-7
196.972
CDJLVNMKXZMCHU-UHFFFAOYSA-N
44886927
B(C1=CC(=CC(=C1)F)C(=O)NC)(O)O
C8H9BFNO3
MFCD07363793
3-fluoro-5-methylcarbamoyl phenylboronic acid, 3-fluoro-5-methylcarbamoyl phenyl boronic acid, 3-fluoro-5-methylcarbamoyl benzeneboronic acid, n-methyl 3-borono-5-fluorobenzamide, 3-fluoro-5-methylaminocarbonyl phenylboronic acid, 3-fluoro-5-methylcarbamoyl phenyl boronic acid, acmc-209qgt, 3-fluoro-5-methylcarbamoyl benzene boronic acid, 3-fluoro-5-methylcarbamoyl-phenyl boronic acid, 3-fluoro-5-methyl carbamoyl phenylboronic acid
[3-fluoro-5-(methylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents