Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-methylbenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-methylbenzyl bromide
CAS 212268-39-8
Molecular Formula C8H8BrF
Formula Weight 203.06
MDL Number MFCD03094316
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2670203

  SDS
alfa aesar™
H2670203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2670206

  SDS
alfa aesar™
H2670206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.06
UN Number UN3265
CAS 212268-39-8
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 1 Danger
MDL Number MFCD03094316
Quantity 1g
Chemical Name or Material 3-Fluoro-5-methylbenzyl bromide
Molecular Formula C8H8BrF
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Applications
3-Fluoro-5-methylbenzyl bromide is used for the production of inorganic bromides. And also used in pharmaceuticals intermediates.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.