Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-methylbenzoic acid, 97%

$138.54 - $415.61

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-methylbenzoic acid
CAS 518070-19-4
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD03094313
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2667703

 
alfa aesar™
H2667703
1g Each for $138.54

AAH2667706

 
alfa aesar™
H2667706
5g Each for $415.61
Description & Specifications

Specifications

Formula Weight 154.14
Chemical Name or Material 3-Fluoro-5-methylbenzoic acid
Assay 97%
Molecular Formula C8H7FO2
MDL Number MFCD03094313
CAS 518070-19-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 141° to 143°C
Quantity 1g

Applications
Intermediate in organic syntheses.

Solubility
Soluble in water (25 deg C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.