missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-methylbenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5311103

 View more versions of this product

Catalog No. AAH5311103



Specifications

Specifications

3-Fluoro-5-methylbenzeneboronic acid
1g
C7H8BFO2
3-fluoro-5-methylphenyl boronic acid, 3-fluoro-5-methylbenzeneboronic acid, 5-fluoro-3-methylphenylboronic acid, 5-fluoro-m-tolylboronic acid, boronic acid,b-3-fluoro-5-methylphenyl, 3-borono-5-fluorotoluene, acmc-209q3i, 3-fluoro-5-methylbenzene boronic acid, 3-fluoranyl-5-methyl-phenyl boronic acid, 3-3-hydroxypyrrolidin-2-yl propanoic acid
B(C1=CC(=CC(=C1)F)C)(O)O
153.947
153.95
216°C to 222°C
850593-06-5
MFCD07363785
QQPLHUGOUZKARP-UHFFFAOYSA-N
(3-fluoro-5-methylphenyl)boronic acid
19261265
98%

Chemical Identifiers

850593-06-5
153.947
QQPLHUGOUZKARP-UHFFFAOYSA-N
19261265
B(C1=CC(=CC(=C1)F)C)(O)O
C7H8BFO2
MFCD07363785
3-fluoro-5-methylphenyl boronic acid, 3-fluoro-5-methylbenzeneboronic acid, 5-fluoro-3-methylphenylboronic acid, 5-fluoro-m-tolylboronic acid, boronic acid,b-3-fluoro-5-methylphenyl, 3-borono-5-fluorotoluene, acmc-209q3i, 3-fluoro-5-methylbenzene boronic acid, 3-fluoranyl-5-methyl-phenyl boronic acid, 3-3-hydroxypyrrolidin-2-yl propanoic acid
(3-fluoro-5-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents