Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-methylbenzaldehyde, 97%

$521.34 - $521.34

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-methylbenzaldehyde
CAS 189628-39-5
Molecular Formula C8H7FO
Formula Weight 138.14
MDL Number MFCD03094315
Synonym 5-Fluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2664806

  SDS
alfa aesar™
H2664806
5g Each for $521.34
Description & Specifications

Specifications

Formula Weight 138.14
Molecular Formula C8H7FO
CAS 189628-39-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
MDL Number MFCD03094315
Health Hazard 1 Warning
Chemical Name or Material 3-Fluoro-5-methylbenzaldehyde
Synonym 5-Fluoro-m-tolualdehyde
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.