missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H53084MD

 View more versions of this product

Catalog No. AAH53084MD



Description

Description

Applications
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid
250mg
C8H8BFO4
3-fluoro-5-methoxycarbonyl phenyl boronic acid, 3-fluoro-5-methoxycarbonylphenyl boronic acid, 3-fluoro-5-methoxycarbonyl phenylboronic acid, 3-fluoro-5-methoxycarbonyl benzeneboronic acid, benzoic acid,3-borono-5-fluoro-, 1-methyl ester 9ci, methyl 3-borono-5-fluorobenzoate, 3-fluoro-5-methoxycarbonyl phenyl boronic acid, acmc-209qg6, ksc657s1f, 3-fluoro-5-methoxycarbonylphenylboronicacid
B(C1=CC(=CC(=C1)F)C(=O)OC)(O)O
197.956
197.96
138°C to 142°C
871329-62-3
MFCD07363749
VKHJVASTGLPBBL-UHFFFAOYSA-N
(3-fluoro-5-methoxycarbonylphenyl)boronic acid
44717206
95%

Chemical Identifiers

871329-62-3
197.956
VKHJVASTGLPBBL-UHFFFAOYSA-N
44717206
B(C1=CC(=CC(=C1)F)C(=O)OC)(O)O
C8H8BFO4
MFCD07363749
3-fluoro-5-methoxycarbonyl phenyl boronic acid, 3-fluoro-5-methoxycarbonylphenyl boronic acid, 3-fluoro-5-methoxycarbonyl phenylboronic acid, 3-fluoro-5-methoxycarbonyl benzeneboronic acid, benzoic acid,3-borono-5-fluoro-, 1-methyl ester 9ci, methyl 3-borono-5-fluorobenzoate, 3-fluoro-5-methoxycarbonyl phenyl boronic acid, acmc-209qg6, ksc657s1f, 3-fluoro-5-methoxycarbonylphenylboronicacid
(3-fluoro-5-methoxycarbonylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents