missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-iodopyridine, 95%

$55.97 - $664.65

Chemical Identifiers

CAS 757950-13-3
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.989
MDL Number MFCD08457780
InChI Key MMJSSIRVEYQWMU-UHFFFAOYSA-N
Synonym 3-fluoro-5-iodo-pyridine, pyridine, 3-fluoro-5-iodo, 3-fluoro-5-iodo-pyridin, pyridine,3-fluoro-5-iodo, 3-fluoranyl-5-iodanyl-pyridine, 3-iodo-5-fluoropyridine, 5-fluoro-3-iodopyridine
PubChem CID 18454778
IUPAC Name 3-fluoro-5-iodopyridine
SMILES C1=C(C=NC=C1I)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64770MD View Documents Alfa Aesar™
H64770MD
250mg Each for $55.97
 
AAH6477003 View Documents Alfa Aesar™
H6477003
1g Each for $183.51
 
AAH6477006 View Documents Alfa Aesar™
H6477006
5g Each for $664.65
 
Specifications

Chemical Identifiers

757950-13-3
222.989
MMJSSIRVEYQWMU-UHFFFAOYSA-N
18454778
C1=C(C=NC=C1I)F
C5H3FIN
MFCD08457780
3-fluoro-5-iodo-pyridine, pyridine, 3-fluoro-5-iodo, 3-fluoro-5-iodo-pyridin, pyridine,3-fluoro-5-iodo, 3-fluoranyl-5-iodanyl-pyridine, 3-iodo-5-fluoropyridine, 5-fluoro-3-iodopyridine
3-fluoro-5-iodopyridine

Specifications

Light Sensitive
250mg
C5H3FIN
11107918
MMJSSIRVEYQWMU-UHFFFAOYSA-N
3-fluoro-5-iodopyridine
18454778
Solid
3-Fluoro-5-iodopyridine
White
757950-13-3
MFCD08457780
3-fluoro-5-iodo-pyridine, pyridine, 3-fluoro-5-iodo, 3-fluoro-5-iodo-pyridin, pyridine,3-fluoro-5-iodo, 3-fluoranyl-5-iodanyl-pyridine, 3-iodo-5-fluoropyridine, 5-fluoro-3-iodopyridine
C1=C(C=NC=C1I)F
222.989
222.99
95%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C