Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-hydroxypyridine, 97%

$165.74 - $468.61

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-hydroxypyridine
CAS 209328-55-2
Molecular Formula C5H4FNO
Formula Weight 113.09
MDL Number MFCD03095199
Synonym 5-Fluoro-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3268303

  SDS
alfa aesar™
H3268303
1g Each for $468.61

AAH32683MD

  SDS
alfa aesar™
H32683MD
250mg Each for $165.74
Description & Specifications

Specifications

Formula Weight 113.09
CAS 209328-55-2
Chemical Name or Material 3-Fluoro-5-hydroxypyridine
MDL Number MFCD03095199
Molecular Formula C5H4FNO
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Synonym 5-Fluoro-3-pyridinol
Quantity 1g
Assay 97%
Health Hazard 1 Warning
Melting Point 161° to 164°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.