missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6223403

 View more versions of this product

Catalog No. AAH6223403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Fluoro-5-hydroxybenzeneboronic acid pinacol ester
1887126-77-3
C12H16BFO3
KBGOBMOAZQOCTA-UHFFFAOYSA-N
3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
72219551
Solid
1g
96%
5-Fluoro-3-hydroxyphenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)O
238.065
238.06

Chemical Identifiers

1887126-77-3
238.065
5-Fluoro-3-hydroxyphenylboronic acid pinacol ester
C12H16BFO3
KBGOBMOAZQOCTA-UHFFFAOYSA-N
72219551
Documents