missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-formylbenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6230303

 View more versions of this product

Catalog No. AAH6230303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Fluoro-5-formylbenzeneboronic acid pinacol ester
1112209-24-1
3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)C=O
250.076
250.07
96%
1g
C13H16BFO3
WENPYTYJEVDZDV-UHFFFAOYSA-N
3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
56776723
Solid

Chemical Identifiers

1112209-24-1
250.076
3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde
3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
C13H16BFO3
WENPYTYJEVDZDV-UHFFFAOYSA-N
56776723
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)C=O
Documents