missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(2-fluorobenzylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5413903

 View more versions of this product

Catalog No. AAH5413903



Specifications

Specifications

1g
2096332-42-0
C14H12BF2NO3
3-Fluoro-5-(2-fluorophenylcarbamoyl)phenylboronic acid
291.06
291.06
3-Fluoro-5-(2-fluorobenzylcarbamoyl)benzeneboronic acid
97%
MFCD20040138
UINFTLMSMWGJLO-UHFFFAOYSA-N
73995653

Chemical Identifiers

2096332-42-0
291.06
UINFTLMSMWGJLO-UHFFFAOYSA-N
73995653
C14H12BF2NO3
MFCD20040138
3-Fluoro-5-(2-fluorophenylcarbamoyl)phenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents