missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54634MD

 View more versions of this product

Catalog No. AAH54634MD



Description

Description

Applications
3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Fluoro-5-(1-pyrrolidinylcarbonyl)benzeneboronic acid
1264616-18-3
MFCD20265305
3-fluoro-5-1-pyrrolidinylcarbonyl benzeneboronic acid
B(C1=CC(=CC(=C1)F)C(=O)N2CCCC2)(O)O
237.037
237.04
250mg
C11H13BFNO3
22139405
XPUNUDDGDYAOFL-UHFFFAOYSA-N
[3-fluoro-5-(pyrrolidine-1-carbonyl)phenyl]boronic acid
73995902
97%

Chemical Identifiers

1264616-18-3
237.037
XPUNUDDGDYAOFL-UHFFFAOYSA-N
73995902
B(C1=CC(=CC(=C1)F)C(=O)N2CCCC2)(O)O
C11H13BFNO3
MFCD20265305
3-fluoro-5-1-pyrrolidinylcarbonyl benzeneboronic acid
[3-fluoro-5-(pyrrolidine-1-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents