Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(trifluoromethyl)pyridine, 98%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)pyridine
CAS 113770-87-9
Molecular Formula C6H3F4N
Formula Weight 165.09
MDL Number MFCD03789250
UN Number UN1993
Synonym alpha,alpha,alpha,3-Tetrafluoropicoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2004303

  SDS
alfa aesar™
L2004303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL20043MD

  SDS
alfa aesar™
L20043MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.09
CAS 113770-87-9
MDL Number MFCD03789250
Synonym alpha,alpha,alpha,3-Tetrafluoropicoline
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 1g
Flash Point 25°C (77°F)
UN Number UN1993
Health Hazard 1 Warning
Molecular Formula C6H3F4N
Assay 98%
Boiling Point 37°C (10mmHg)
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)pyridine
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.