Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Fluoro-4'-(trifluoromethyl)propiophenone, 97+%

$94.93 - $336.01

Chemical Identifiers

Chemical Name or Material 3'-Fluoro-4'-(trifluoromethyl)propiophenone
CAS 237761-78-3
Molecular Formula C10H8F4O
Formula Weight 220.17
MDL Number MFCD00236306
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2281403

  SDS
alfa aesar™
B2281403
1g Each for $94.93

AAB2281406

  SDS
alfa aesar™
B2281406
5g Each for $336.01
Description & Specifications

Specifications

Formula Weight 220.17
Assay ≥97%
MDL Number MFCD00236306
CAS 237761-78-3
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Quantity 1g
Chemical Name or Material 3'-Fluoro-4'-(trifluoromethyl)propiophenone
Molecular Formula C10H8F4O
Melting Point 35° to 38°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.