Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoyl chloride
CAS 216144-68-2
Molecular Formula C8H3ClF4O
Formula Weight 226.56
MDL Number MFCD00236284
UN Number UN3265
Synonym alpha,alpha,alpha,4-Tetrafluoro-p-toluoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2045403

  SDS
alfa aesar™
B2045403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2045406

  SDS
alfa aesar™
B2045406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.56
Quantity 1g
Molecular Formula C8H3ClF4O
MDL Number MFCD00236284
Flash Point 81°C (177°F)
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Boiling Point 151° to 153°C
Assay 97%
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoyl chloride
CAS 216144-68-2
Synonym alpha,alpha,alpha,4-Tetrafluoro-p-toluoyl chloride
Health Hazard 1 Danger
UN Number UN3265
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and active metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.