Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%

$83.18 - $319.43

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzaldehyde
CAS 204339-72-0
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD00236275
Synonym alpha,alpha,alpha,3-Tetrafluoro-p-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2000803

  SDS
alfa aesar™
B2000803
1g Each for $83.18

AAB2000806

  SDS
alfa aesar™
B2000806
5g Each for $319.43
Description & Specifications

Specifications

Formula Weight 192.11
Quantity 1g
CAS 204339-72-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 73°C (163°F)
Molecular Formula C8H4F4O
Assay 97%
Synonym alpha,alpha,alpha,3-Tetrafluoro-p-tolualdehyde
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzaldehyde
MDL Number MFCD00236275

Applications
3-Fluoro-4-(trifluoromethyl)benzaldehyde is used as pharmaceutical intermediates.

Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.