missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(trifluoromethoxy)phenylacetic acid, 97%

$514.59 - $514.59

Chemical Identifiers

Molecular Formula C9H6F4O3
Formula Weight 238.14
MDL Number MFCD06660343
Chemical Name or Material 3-Fluoro-4-(trifluoromethoxy)phenylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2681603 SDS Alfa Aesar™
H2681603
1g Each for $514.59
AAH26816MD SDS Alfa Aesar™
H26816MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C9H6F4O3
238.14
MFCD06660343
3-Fluoro-4-(trifluoromethoxy)phenylacetic acid

Specifications

1g
238.14
3-Fluoro-4-(trifluoromethoxy)phenylacetic acid
C9H6F4O3
MFCD06660343
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning