missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(trifluoromethoxy)benzoic acid, 97%

$210.88 - $210.88

Chemical Identifiers

CAS 886498-89-1
Molecular Formula C8H4F4O3
Formula Weight 224.11
MDL Number MFCD06660202
Chemical Name or Material 3-Fluoro-4-(trifluoromethoxy)benzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2623403 SDS Alfa Aesar™
H2623403
1g Each for $210.88
AAH2623406 SDS Alfa Aesar™
H2623406
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Description & Specifications

Chemical Identifiers

886498-89-1
224.11
3-Fluoro-4-(trifluoromethoxy)benzoic acid
C8H4F4O3
MFCD06660202

Specifications

98° to 101°C
886498-89-1
224.11
3-Fluoro-4-(trifluoromethoxy)benzoic acid
1g
C8H4F4O3
MFCD06660202
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning