Please login to your online account to display your discounted pricing

3-Fluoro-4-nitrotoluene, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-nitrotoluene
CAS 446-34-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H6FNO2
Linear Formula CH3C6H3(NO2)F
Formula Weight 155.13
MDL Number MFCD00007053
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC255750050

  SDS
thermo fisher scientific
255750050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystals or Needles
Formula Weight 155.13
Quantity 5g
Color Light yellow to light brown
Melting Point 52° to 55°C
Beilstein 05, 251
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C7H6FNO2
Chemical Name or Material 3-Fluoro-4-nitrotoluene
CAS 446-34-4
Assay Percent Range 99+%
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00007053
Linear Formula CH3C6H3(NO2)F
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.