Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-nitrobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-nitrobenzeneboronic acid pinacol ester
CAS 939968-60-2
Formula Weight 267.06
Synonym 3-Fluoro-4-nitrophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6292103

 
alfa aesar™
H6292103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62921MD

 
alfa aesar™
H62921MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267.06
Physical Form Solid
Quantity 1g
Assay 96%
CAS 939968-60-2
Synonym 3-Fluoro-4-nitrophenylboronic acid pinacol ester
Chemical Name or Material 3-Fluoro-4-nitrobenzeneboronic acid pinacol ester