Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-nitroaniline, 97%

$159.72 - $403.81

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-nitroaniline
CAS 2369-13-3
Molecular Formula C6H5FN2O2
Formula Weight 156.12
MDL Number MFCD03412239
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2889503

  SDS
alfa aesar™
H2889503
1g Each for $403.81

AAH28895MD

  SDS
alfa aesar™
H28895MD
250mg Each for $159.72
Description & Specifications

Specifications

Formula Weight 156.12
Quantity 1g
Molecular Formula C6H5FN2O2
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 158° to 162°C
Assay 97%
Health Hazard 1 Danger
Chemical Name or Material 3-Fluoro-4-nitroaniline
CAS 2369-13-3
MDL Number MFCD03412239
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.