missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4'-n-propylbiphenyl-4-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H64071MD

 View more versions of this product

Catalog No. AAH64071MD



Description

Description

Applications
3-Fluoro-4′-n-propylbiphenyl-4-boronic acid is used in OLED.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

3-Fluoro-4'-n-propylbiphenyl-4-boronic acid
909709-42-8
MFCD10687198
3-fluoro-4'-propyl-1,1'-biphenyl-4-yl boronic acid, 4'-propyl-3-fluorobiphenyl-4-boronic acid, 4-propyl-3-fluorobiphenyl-4-boronic acid, 3-fluoro-4'-propyl-biphenylboronic acid, 3-fluoro-4'-propyl-4-biphenylboronic acid, 4'-propyl-3-fluoro-4-biphenyl boronic acid, 2-fluoro-4-4-propylphenyl phenylboronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl
B(C1=C(C=C(C=C1)C2=CC=C(C=C2)CCC)F)(O)O
258.099
258.1
98%
250mg
C15H16BFO2
18339837
XZGCBSYWWZAXRC-UHFFFAOYSA-N
[2-fluoro-4-(4-propylphenyl)phenyl]boronic acid
53402576
Solid

Chemical Identifiers

909709-42-8
258.099
XZGCBSYWWZAXRC-UHFFFAOYSA-N
53402576
B(C1=C(C=C(C=C1)C2=CC=C(C=C2)CCC)F)(O)O
C15H16BFO2
MFCD10687198
3-fluoro-4'-propyl-1,1'-biphenyl-4-yl boronic acid, 4'-propyl-3-fluorobiphenyl-4-boronic acid, 4-propyl-3-fluorobiphenyl-4-boronic acid, 3-fluoro-4'-propyl-biphenylboronic acid, 3-fluoro-4'-propyl-4-biphenylboronic acid, 4'-propyl-3-fluoro-4-biphenyl boronic acid, 2-fluoro-4-4-propylphenyl phenylboronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl boronic acid, boronic acid, 3-fluoro-4'-propyl 1,1'-biphenyl-4-yl
[2-fluoro-4-(4-propylphenyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents