missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Fluoro-4-n-octyloxybenzoic Acid 97.0+%, TCI America™
$469.49 - $1406.83
Chemical Identifiers
CAS | 326-78-3 |
---|---|
Molecular Formula | C15H21FO3 |
Molecular Weight (g/mol) | 268.328 |
MDL Number | MFCD00191649 |
InChI Key | SQVFTDZEUGEJTQ-UHFFFAOYSA-N |
PubChem CID | 593523 |
IUPAC Name | 3-fluoro-4-octoxybenzoic acid |
SMILES | CCCCCCCCOC1=C(C=C(C=C1)C(=O)O)F |
Chemical Identifiers
326-78-3 | |
268.328 | |
SQVFTDZEUGEJTQ-UHFFFAOYSA-N | |
3-fluoro-4-octoxybenzoic acid |
C15H21FO3 | |
MFCD00191649 | |
593523 | |
CCCCCCCCOC1=C(C=C(C=C1)C(=O)O)F |
Specifications
326-78-3 | |
C15H21FO3 | |
1 g | |
CCCCCCCCOC1=C(C=C(C=C1)C(=O)O)F | |
268.328 | |
268.33 | |
Crystalline Powder |
White-Yellow | |
MFCD00191649 | |
SQVFTDZEUGEJTQ-UHFFFAOYSA-N | |
3-fluoro-4-octoxybenzoic acid | |
593523 | |
≥97.0% (GC,T) | |
3-Fluoro-4-n-octyloxybenzoic Acid |
Safety and Handling
TSCA : No