Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methylpyridine, 95%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methylpyridine
CAS 399-88-2
Formula Weight 111.12
MDL Number MFCD04114226
UN Number UN1993
Beilstein 107788
Synonym 3-Fluoro-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6474703

  SDS
alfa aesar™
H6474703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6474706

  SDS
alfa aesar™
H6474706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 111.12
Color Colorless to pale yellow
Beilstein 107788
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 136°C (748mmHg)
Refractive Index 1.4795
Chemical Name or Material 3-Fluoro-4-methylpyridine
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
UN Number UN1993
CAS 399-88-2
Density 1.08g/mL
Health Hazard 1 Warning
MDL Number MFCD04114226
Physical Form Liquid
Synonym 3-Fluoro-4-picoline
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.