Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methylphenylacetic acid, 97%

$89.80 - $221.50

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methylphenylacetic acid
CAS 261951-74-0
Molecular Formula C9H9FO2
Formula Weight 168.17
MDL Number MFCD01631536
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2669003

  SDS
alfa aesar™
H2669003
1g Each for $221.50

AAH26690MD

  SDS
alfa aesar™
H26690MD
250mg Each for $89.80
Description & Specifications

Specifications

Formula Weight 168.17
Assay 97%
Molecular Formula C9H9FO2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-Fluoro-4-methylphenylacetic acid
MDL Number MFCD01631536
CAS 261951-74-0
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 104° to 107°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.