missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5328903

 View more versions of this product

Catalog No. AAH5328903



Specifications

Specifications

3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid
1g
C8H9BFNO3
3-fluoro-4-methylcarbamoyl phenyl boronic acid, n-methyl 4-borono-2-fluorobenzamide, 3-fluoro-4-methylcarbamoyl phenylboronic acid, 3-fluoro-4-methylcarbamoyl benzeneboronic acid, 3-fluoro-4-methylcarbamoyl phenyl boronic acid, acmc-209pz8, synquest 6660-3-t7, n-methyl-4-borono-2-fluorobenzamide, 3-fluoro-4-methylcarbamoyl phenyl boronicacid, 3-fluoranyl-4-methylcarbamoyl phenyl boronic acid
B(C1=CC(=C(C=C1)C(=O)NC)F)(O)O
196.972
196.97
173°C to 175°C
849833-86-9
MFCD08436026
IVSXSBGNMOZTJR-UHFFFAOYSA-N
[3-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
44717467
98%

Chemical Identifiers

849833-86-9
196.972
IVSXSBGNMOZTJR-UHFFFAOYSA-N
44717467
B(C1=CC(=C(C=C1)C(=O)NC)F)(O)O
C8H9BFNO3
MFCD08436026
3-fluoro-4-methylcarbamoyl phenyl boronic acid, n-methyl 4-borono-2-fluorobenzamide, 3-fluoro-4-methylcarbamoyl phenylboronic acid, 3-fluoro-4-methylcarbamoyl benzeneboronic acid, 3-fluoro-4-methylcarbamoyl phenyl boronic acid, acmc-209pz8, synquest 6660-3-t7, n-methyl-4-borono-2-fluorobenzamide, 3-fluoro-4-methylcarbamoyl phenyl boronicacid, 3-fluoranyl-4-methylcarbamoyl phenyl boronic acid
[3-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents