Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methylbenzyl alcohol, 97%

$49.11 - $130.47

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methylbenzyl alcohol
CAS 192702-79-7
Molecular Formula C8H9FO
Formula Weight 140.16
MDL Number MFCD01631533
Beilstein 7700178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1917003

  SDS
alfa aesar™
L1917003
1g Each for $130.47

AAL19170MD

  SDS
alfa aesar™
L19170MD
250mg Each for $49.11
Description & Specifications

Specifications

Formula Weight 140.16
CAS 192702-79-7
MDL Number MFCD01631533
Chemical Name or Material 3-Fluoro-4-methylbenzyl alcohol
Assay 97%
Molecular Formula C8H9FO
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 7700178
Melting Point 33° to 36°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.