missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-4-methylbenzoyl chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 59189-97-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H6ClFO
Linear Formula CH3C6H3(F)C(O)Cl
Formula Weight 172.59
MDL Number MFCD00671774
Chemical Name or Material 3-Fluoro-4-methylbenzoyl chloride
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC375800010 SDS Thermo Fisher Scientific
AC375800010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

59189-97-8
100.0
C8H6ClFO
172.59
3-Fluoro-4-methylbenzoyl chloride
96.0
97%
CH3C6H3(F)C(O)Cl
MFCD00671774
≥96.0%

Specifications

172.59
1g
100.0
C8H6ClFO
Authentic
1.5320 to 1.5340 (20°C, 589nm)
MFCD00671774
3-Fluoro-4-methylbenzoyl chloride
Liquid
Clear yellow
96.0
59189-97-8
Glass bottle
CH3C6H3(F)C(O)Cl
97%
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger