Please login to your online account to display your discounted pricing

3-Fluoro-4-methylbenzoic acid, 97%, ACROS Organics™

$92.99 - $536.57

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methylbenzoic acid
Name Note 97%
CAS 350-28-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)CO2H
Formula Weight 154.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119470025

  SDS
thermo fisher scientific
119470025
Glass bottle 2.5g Each for $92.99

AC119470250

  SDS
thermo fisher scientific
AC119470250
Glass bottle 25g Each for $536.57
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 154.14
Color White to light grayish-beige
Quantity 2.5g
Beilstein 09, 497
Melting Point 170° to 175°C
CAS Max % 100.0
CAS Min % 96.0
Assay Percent Range 97%
Molecular Formula C8H7FO2
Linear Formula FC6H3(CH3)CO2H
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Chemical Name or Material 3-Fluoro-4-methylbenzoic acid
Name Note 97%
CAS 350-28-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.