Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methylbenzaldehyde, 97%

$123.24 - $123.24

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methylbenzaldehyde
CAS 177756-62-6
Molecular Formula C8H7FO
Formula Weight 138.14
MDL Number MFCD01631532
Beilstein 7699714
Synonym 3-Fluoro-p-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2373606

  SDS
alfa aesar™
B2373606
5g Each for $123.24
Description & Specifications

Specifications

Formula Weight 138.14
Quantity 5g
Boiling Point 206°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material 3-Fluoro-4-methylbenzaldehyde
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 7699714
Molecular Formula C8H7FO
CAS 177756-62-6
MDL Number MFCD01631532
Assay 97%
Synonym 3-Fluoro-p-tolualdehyde
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.