Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methylaniline, 98+%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methylaniline
CAS 452-77-7
Molecular Formula C7H8FN
Formula Weight 125.15
MDL Number MFCD00007762
UN Number UN1759
Beilstein 2715987
Synonym 4-Amino-2-fluorotoluene; 3-Fluoro-p-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1448414

  SDS
alfa aesar™
A1448414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1448422

  SDS
alfa aesar™
A1448422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 125.15
Health Hazard 2 GHS H Statement
H314-H373-H302-H312-H332
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Molecular Formula C7H8FN
Assay ≥98%
Odor Characteristic
MDL Number MFCD00007762
Quantity 25g
Melting Point 30° to 35°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
UN Number UN1759
Boiling Point 93°C (12mmHg)
Beilstein 2715987
CAS 452-77-7
RTECS Number XU6825000
Synonym 4-Amino-2-fluorotoluene; 3-Fluoro-p-toluidine
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material 3-Fluoro-4-methylaniline
Flash Point 88°C (190°F)
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H373-H302-H312-H332
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.