Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methyl-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methyl-DL-phenylalanine
Molecular Formula C10H12FNO2
Formula Weight 197.21
MDL Number MFCD09832258
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3219403

 
alfa aesar™
H3219403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32194MD

 
alfa aesar™
H32194MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.21
Chemical Name or Material 3-Fluoro-4-methyl-DL-phenylalanine
Assay 97%
Molecular Formula C10H12FNO2
MDL Number MFCD09832258
Melting Point ≈368°C
Quantity 1g