missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methoxypyridine, 98%

$48.58 - $647.69

Chemical Identifiers

CAS 1060805-03-9
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD13185605
InChI Key XYTUUTJNWFECNS-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxy-pyridine, pyridine, 3-fluoro-4-methoxy, abbypharma ap-14-5665
PubChem CID 57336639
IUPAC Name 3-fluoro-4-methoxypyridine
SMILES COC1=C(C=NC=C1)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64603MD View Documents Alfa Aesar™
H64603MD
250mg Each for $48.58
 
AAH6460303 View Documents Alfa Aesar™
H6460303
1g Each for $151.12
 
AAH6460306 View Documents Alfa Aesar™
H6460306
5g Each for $647.69
 
Specifications

Chemical Identifiers

1060805-03-9
127.118
XYTUUTJNWFECNS-UHFFFAOYSA-N
57336639
COC1=C(C=NC=C1)F
C6H6FNO
MFCD13185605
3-fluoro-4-methoxy-pyridine, pyridine, 3-fluoro-4-methoxy, abbypharma ap-14-5665
3-fluoro-4-methoxypyridine

Specifications

250mg
C6H6FNO
19561531
XYTUUTJNWFECNS-UHFFFAOYSA-N
3-fluoro-4-methoxypyridine
57336639
98%
1060805-03-9
MFCD13185605
3-fluoro-4-methoxy-pyridine, pyridine, 3-fluoro-4-methoxy, abbypharma ap-14-5665
COC1=C(C=NC=C1)F
127.118
127.12
3-Fluoro-4-methoxypyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning