Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methoxybenzylamine, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methoxybenzylamine
CAS 123652-95-9
Molecular Formula C8H10FNO
Formula Weight 155.17
MDL Number MFCD04116360
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2660703

  SDS
alfa aesar™
H2660703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2660709

  SDS
alfa aesar™
H2660709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.17
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 1 Danger
Chemical Name or Material 3-Fluoro-4-methoxybenzylamine
Molecular Formula C8H10FNO
Quantity 1g
Assay 97%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD04116360
UN Number UN2735
CAS 123652-95-9
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

Applications
3-Fluoro-4-methoxybenzylamine is used as an intermediate in organic synthesis.

Solubility
Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with oxidizing agents, bases, acids and carbon dioxide.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.