Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-methoxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methoxybenzeneboronic acid pinacol ester
CAS 754226-34-1
Formula Weight 252.09
MDL Number MFCD05863912
Synonym 3-Fluoro-4-methoxyphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6298503

 
alfa aesar™
H6298503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62985MD

 
alfa aesar™
H62985MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.09
CAS 754226-34-1
Physical Form Solid
MDL Number MFCD05863912
Assay 96%
Quantity 1g
Synonym 3-Fluoro-4-methoxyphenylboronic acid pinacol ester
Chemical Name or Material 3-Fluoro-4-methoxybenzeneboronic acid pinacol ester