Please login to your online account to display your discounted pricing

3-Fluoro-4-methoxybenzaldehyde, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-methoxybenzaldehyde
CAS 351-54-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7FO2
Linear Formula FC6H3(OCH3)CHO
Formula Weight 154.14
MDL Number MFCD00003349
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC168210010

  SDS
thermo fisher scientific
AC168210010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC168210050

  SDS
thermo fisher scientific
AC168210050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Low Melting Crystalline Mass
Formula Weight 154.14
Quantity 1g
Color White to light yellow
Melting Point 30° to 35°C
Density 113g/mL
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzaldehyde
CAS 351-54-2
Molecular Formula C8H7FO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00003349
CAS Min % 97.5
Linear Formula FC6H3(OCH3)CHO
Synonym 3-Fluoro-p-anisaldehyde
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.