Please login to your online account to display your discounted pricing

3-Fluoro-4-methoxybenzaldehyde, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC168210050

 View more versions of this product

Catalog No. AC168210050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Low Melting Crystalline Mass
Assay ≥97.5%
Formula Weight 154.14
Quantity 5g
Color White to light yellow
Melting Point 30° to 35°C
Density 113g/mL
CAS Max % 100.0
Packaging Glass Bottle
Assay Percent Range 98%
CAS 351-54-2
Linear Formula FC6H3(OCH3)CHO
Chemical Name or Material 3-Fluoro-4-methoxybenzaldehyde
Molecular Formula C8H7FO2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
MDL Number MFCD00003349
CAS Min % 97.5
Infrared Spectrum Authentic
Synonym 3-Fluoro-p-anisaldehyde

Specifications

Chemical Name or Material 3-Fluoro-4-methoxybenzaldehyde
CAS 351-54-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7FO2
Linear Formula FC6H3(OCH3)CHO
Formula Weight 154.14
MDL Number MFCD00003349
Synonym 3-Fluoro-p-anisaldehyde
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

SDS & Safety Documents