missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-4-methoxybenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC168210010

 View more versions of this product

Catalog No. AC168210010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
154.14
White to light yellow
113g/mL
Glass bottle
98%
351-54-2
MFCD00003349
FC6H3(OCH3)CHO
Low Melting Crystalline Mass
1g
30° to 35°C
100.0
Authentic
3-Fluoro-4-methoxybenzaldehyde
C8H7FO2
97.5
3-Fluoro-p-anisaldehyde

Chemical Identifiers

351-54-2
100.0
C8H7FO2
154.14
3-Fluoro-p-anisaldehyde
≥97.5%
97.5
98%
FC6H3(OCH3)CHO
MFCD00003349
3-Fluoro-4-methoxybenzaldehyde

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.