missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-iodotoluene, 97%

Manufacturer:  Alfa Aesar™ H5349106

 View more versions of this product

Catalog No. AAH5349106



Specifications

Specifications

3-Fluoro-4-iodotoluene
92°C (198°F)
Light sensitive
5g
C7H6FI
2-fluoro-1-iodo-4-methyl-benzene
CC1=CC(=C(C=C1)I)F
236.028
236.03
1.803
1.581
138°C to 139°C (9mmHg)
452-79-9
MFCD00143162
XVHYSMVSLDIUCM-UHFFFAOYSA-N
2-fluoro-1-iodo-4-methylbenzene
2774524
97%

Chemical Identifiers

452-79-9
236.028
XVHYSMVSLDIUCM-UHFFFAOYSA-N
2774524
CC1=CC(=C(C=C1)I)F
C7H6FI
MFCD00143162
2-fluoro-1-iodo-4-methyl-benzene
2-fluoro-1-iodo-4-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

TSCA : No

Recommended Storage : Ambient temperatures

Documents