Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-iodopyridine, 98%

$209.34 - $878.93

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-iodopyridine
CAS 22282-75-3
Molecular Formula C5H3FIN
Formula Weight 222.99
MDL Number MFCD03092927
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5006803

 
alfa aesar™
H5006803
1g Each for $209.34

AAH5006806

 
alfa aesar™
H5006806
5g Each for $878.93
Description & Specifications

Specifications

Formula Weight 222.99
Chemical Name or Material 3-Fluoro-4-iodopyridine
Assay 98%
Molecular Formula C5H3FIN
MDL Number MFCD03092927
CAS 22282-75-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 87° to 90°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.