missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-iodobenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H55141MD

 View more versions of this product

Catalog No. AAH55141MD



Specifications

Specifications

3-Fluoro-4-iodobenzonitrile
250mg
C7H3FIN
3-fluoro-4-iodo benzonitrile
C1=CC(=C(C=C1C#N)F)I
247.011
247.01
113°C to 117°C
887266-99-1
MFCD07782076
NPKQMCCUPXZXFI-UHFFFAOYSA-N
3-fluoro-4-iodobenzonitrile
24721521
97%

Chemical Identifiers

887266-99-1
247.011
NPKQMCCUPXZXFI-UHFFFAOYSA-N
24721521
C1=CC(=C(C=C1C#N)F)I
C7H3FIN
MFCD07782076
3-fluoro-4-iodo benzonitrile
3-fluoro-4-iodobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents