missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-iodoaniline, 98%

Manufacturer:  Alfa Aesar™ H61427MD

 View more versions of this product

Catalog No. AAH61427MD



Description

Description

Applications
3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water (228.3 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Specifications

3-Fluoro-4-iodoaniline
250mg
C6H5FIN
UN2811
Slightly soluble in water (228.3mg/L at 25°C).
C1=CC(=C(C=C1N)F)I
237.016
237.01
98%
Light sensitive
656-66-6
MFCD07774187
3-fluoro-4-iodo-aniline, 3-fluoro-4-iodo-phenylamine, 3-fluoro-4-iodophenylamine, 4-iodo 3-fluoro aniline, acmc-1b9hq, benzenamine, 3-fluoro-4-iodo, buttpark 120\07-56, pubchem1534, zlchem 541
KUVVJHBHRIXJKI-UHFFFAOYSA-N
3-fluoro-4-iodoaniline
15475546
Solid

Chemical Identifiers

656-66-6
237.016
KUVVJHBHRIXJKI-UHFFFAOYSA-N
15475546
C1=CC(=C(C=C1N)F)I
C6H5FIN
MFCD07774187
3-fluoro-4-iodo-aniline, 3-fluoro-4-iodo-phenylamine, 3-fluoro-4-iodophenylamine, 4-iodo 3-fluoro aniline, acmc-1b9hq, benzenamine, 3-fluoro-4-iodo, buttpark 120\07-56, pubchem1534, zlchem 541
3-fluoro-4-iodoaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications