missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-iodoaniline, 98%

Manufacturer:  Alfa Aesar™ H6142706

 View more versions of this product

Catalog No. AAH6142706



Description

Description

Applications
3-Fluoro-4-iodoaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water (228.3 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Specifications

3-Fluoro-4-iodoaniline
5g
C6H5FIN
UN2811
KUVVJHBHRIXJKI-UHFFFAOYSA-N
3-fluoro-4-iodoaniline
15475546
Solid
Light Sensitive
656-66-6
MFCD07774187
3-fluoro-4-iodophenylamine, benzenamine, 3-fluoro-4-iodo, 3-fluoro-4-iodo-aniline, zlchem 541, pubchem1534, 4-iodo 3-fluoro aniline, acmc-1b9hq, 3-fluoro-4-iodo-phenylamine, buttpark 120\07-56
C1=CC(=C(C=C1N)F)I
237.016
237.01
98%

Chemical Identifiers

656-66-6
237.016
KUVVJHBHRIXJKI-UHFFFAOYSA-N
15475546
C1=CC(=C(C=C1N)F)I
C6H5FIN
MFCD07774187
3-fluoro-4-iodophenylamine, benzenamine, 3-fluoro-4-iodo, 3-fluoro-4-iodo-aniline, zlchem 541, pubchem1534, 4-iodo 3-fluoro aniline, acmc-1b9hq, 3-fluoro-4-iodo-phenylamine, buttpark 120\07-56
3-fluoro-4-iodoaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents