missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
3-Fluoro-4-hydroxyphenylacetic acid, 98%, ACROS Organics™
Chemical Identifiers
CAS
458-09-3
Molecular Formula
C8H7FO3
Molecular Weight (g/mol)
170.139
MDL Number
MFCD00004348
InChI Key
YRFBZAHYMOSSGX-UHFFFAOYSA-N
Synonym
2-3-fluoro-4-hydroxy-phenyl acetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxy-phenyl-acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenylacetic acid, 4-carboxymethyl-2-fluorophenol, acmc-1ahjh, benzeneacetic acid, 3-fluoro-4-hydroxy, opera_id_1839
PubChem CID
68014
IUPAC Name
2-(3-fluoro-4-hydroxyphenyl)acetic acid
SMILES
C1=CC(=C(C=C1CC(=O)O)F)O
View More Specs
Products
1
Catalog Number
Mfr. No.
Quantity
Packaging
Price
Quantity
AC208560050
SDS
View Documents
Thermo Fisher Scientific
AC208560050
5g
Glass bottle
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Specifications
Chemical Identifiers
CAS
458-09-3
Molecular Weight (g/mol)
170.139
InChI Key
YRFBZAHYMOSSGX-UHFFFAOYSA-N
PubChem CID
68014
SMILES
C1=CC(=C(C=C1CC(=O)O)F)O
Molecular Formula
C8H7FO3
MDL Number
MFCD00004348
Synonym
2-3-fluoro-4-hydroxy-phenyl acetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxy-phenyl-acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenylacetic acid, 4-carboxymethyl-2-fluorophenol, acmc-1ahjh, benzeneacetic acid, 3-fluoro-4-hydroxy, opera_id_1839
IUPAC Name
2-(3-fluoro-4-hydroxyphenyl)acetic acid
Specifications
Infrared Spectrum
Authentic
Color
White to Beige
Quantity
5g
CAS Min %
97.5
Assay Percent Range
98%
Linear Formula
FC6 H3 (OH)CH2 CO2 H
Beilstein
10, III, 440
Synonym
2-3-fluoro-4-hydroxy-phenyl acetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxy-phenyl-acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenylacetic acid, 4-carboxymethyl-2-fluorophenol, acmc-1ahjh, benzeneacetic acid, 3-fluoro-4-hydroxy, opera_id_1839
SMILES
C1=CC(=C(C=C1CC(=O)O)F)O
Molecular Weight (g/mol)
170.139
Formula Weight
170.14
Percent Purity
98%
Packaging
Glass bottle
Melting Point
132.0°C to 134.0°C
CAS
458-09-3
CAS Max %
100.0
Molecular Formula
C8H7FO3
MDL Number
MFCD00004348
Merck Index
10, 4079
InChI Key
YRFBZAHYMOSSGX-UHFFFAOYSA-N
IUPAC Name
2-(3-fluoro-4-hydroxyphenyl)acetic acid
PubChem CID
68014
Physical Form
Crystalline Powder
Chemical Name or Material
3-Fluoro-4-hydroxyphenylacetic acid
Safety and Handling
Safety and Handling
GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
SDS