Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-hydroxybenzoic acid, 98%

$99.30 - $360.12

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-hydroxybenzoic acid
CAS 350-29-8
Molecular Formula C7H5FO3
Formula Weight 156.11
MDL Number MFCD00143161
Beilstein 2829823
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2216003

  SDS
alfa aesar™
B2216003
1g Each for $99.30

AAB2216006

  SDS
alfa aesar™
B2216006
5g Each for $360.12
Description & Specifications

Specifications

Formula Weight 156.11
Melting Point 157° to 159°C (dec.)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Beilstein 2829823
Molecular Formula C7H5FO3
MDL Number MFCD00143161
Assay 98%
CAS 350-29-8
Health Hazard 1 Warning
Chemical Name or Material 3-Fluoro-4-hydroxybenzoic acid

Applications
3-fluoro-4- hydroxybenzoic acid is the most potent protocatechuate 3,4-dioxygenase inhibitor.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.