Please login to your online account to display your discounted pricing

3-Fluoro-4-hydroxybenzoic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Fluoro-4-hydroxybenzoic acid
CAS 350-29-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C7H5FO3
Formula Weight 156.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441890010

 
thermo fisher scientific
AC441890010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441890050

 
thermo fisher scientific
AC441890050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Assay ≥94.0%
Formula Weight 156.11
Quantity 1g
Color Off-white to brown
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 3-Fluoro-4-hydroxybenzoic acid
Assay Percent Range 95%
Molecular Formula C7H5FO3
CAS 350-29-8
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.