missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-4-hydroxybenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H53110MD

 View more versions of this product

Catalog No. AAH53110MD



Description

Description

Applications
3-Fluoro-4-hydroxybenzeneboronic acid is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

3-Fluoro-4-hydroxybenzeneboronic acid
250mg
C6H6BFO3
3-fluoro-4-hydroxyphenyl boronic acid, 4-borono-2-fluorophenol, boronic acid, 3-fluoro-4-hydroxyphenyl, 3-fluoro-4-hydroxybenzeneboronic acid, pubchem18807, acmc-1c2s7, 3-fluoro-4-hydroxy-phenylboronic acid, 3-fluoro-4-hydroxyphenyl boronic acid, 4-hydroxy-3-fluorophenyl boronic acid, 3-fluoro-4-hydroxyphenyl-boronic acid
B(C1=CC(=C(C=C1)O)F)(O)O
155.919
155.92
252°C to 256°C
182344-14-5
MFCD06659838
OYNDLOJPYURCJG-UHFFFAOYSA-N
(3-fluoro-4-hydroxyphenyl)boronic acid
23005361
97%

Chemical Identifiers

182344-14-5
155.919
OYNDLOJPYURCJG-UHFFFAOYSA-N
23005361
B(C1=CC(=C(C=C1)O)F)(O)O
C6H6BFO3
MFCD06659838
3-fluoro-4-hydroxyphenyl boronic acid, 4-borono-2-fluorophenol, boronic acid, 3-fluoro-4-hydroxyphenyl, 3-fluoro-4-hydroxybenzeneboronic acid, pubchem18807, acmc-1c2s7, 3-fluoro-4-hydroxy-phenylboronic acid, 3-fluoro-4-hydroxyphenyl boronic acid, 4-hydroxy-3-fluorophenyl boronic acid, 3-fluoro-4-hydroxyphenyl-boronic acid
(3-fluoro-4-hydroxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents