Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Fluoro-4'-hydroxyacetophenone, 98+%

$86.19 - $245.61

Chemical Identifiers

Chemical Name or Material 3'-Fluoro-4'-hydroxyacetophenone
CAS 403-14-5
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD00215840
Beilstein 1936529
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1989203

  SDS
alfa aesar™
L1989203
1g Each for $245.61

AAL19892MD

  SDS
alfa aesar™
L19892MD
250mg Each for $86.19
Description & Specifications

Specifications

Formula Weight 154.14
Chemical Name or Material 3'-Fluoro-4'-hydroxyacetophenone
Assay ≥98%
MDL Number MFCD00215840
CAS 403-14-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Beilstein 1936529
Quantity 1g
Melting Point 127° to 129°C
Molecular Formula C8H7FO2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.